missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, Puratronic™, 99.997% (metals basis)

Catalog No. AA1070918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1070918 50 g
AA1070909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1070918 Supplier Thermo Scientific Chemicals Supplier No. 01070918

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850°C (volat.)
Physical Form Powder
Quantity 50 g
Merck Index 14,4952
Formula Weight 277.64
Percent Purity 99.997%
Grade Puratronic
Odor Odorless
Density 7.18 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) oxide
Show More Show Less
EINECSNumber 215-193-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only