missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, 99.995% (metals basis)

Catalog No. p-4824756
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Catalog No. Quantity
AA4546009 10 g
AA4546018 50 g
2 options

Catalog No. AA4546009

Supplier: Thermo Scientific Chemicals 04546009

Only null left

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

Indium oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850° volat.
Color Yellow
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Insoluble in water and soluble in acids.
Formula Weight 277.64
Odor Odorless
Density 7.18
Chemical Name or Material Indium(III) oxide
EINECSNumber 215-193-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only