missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1070806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1070806 5 g
AA1070814 25 g
AA1070822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1070806 Supplier Thermo Scientific Chemicals Supplier No. 01070806

CAS: 207398-97-8 | InN3O9 | 300.83 g/mol

Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207398-97-8
Molecular Formula InN3O9
Molecular Weight (g/mol) 300.83
MDL Number MFCD00149742
InChI Key LKRFCKCBYVZXTC-UHFFFAOYSA-N
Synonym indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2o
PubChem CID 3084148
SMILES [In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 100°C (in -H2O)
Color White
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1477
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name indium(3+) trinitrate
Formula Weight 300.83 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Indium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 237-393-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only