missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol

Catalog No. aa44562ac
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44562AC 25 mL
AA44562AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44562AC Supplier Thermo Scientific Chemicals Supplier No. 044562AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol

Precursor used in the preparation of indium oxide containing films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38218-24-5
Molecular Formula C9H21InO3
Molecular Weight (g/mol) 292.082
MDL Number MFCD00210318
InChI Key OVZUSPADPSOQQN-UHFFFAOYSA-N
Synonym triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
PubChem CID 57346963
IUPAC Name indium(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]

Specifications

Color Colorless
Density 0.78 g/mL
Boiling Point 82°C
Flash Point 12°C (54°F)
Linear Formula In[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ketones and esters.
Formula Weight 292.08
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 5% w/v isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Indium (III) isopropoxide
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only