missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) isopropoxide, 99.9% (metals basis)

Precursor used in the preparation of indium oxide containing films | CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol

Supplier:  Thermo Scientific Chemicals 04209804

 View more versions of this product

Catalog No. AA4209804


Only null left
Explore available promotions

Description

Description

Indium(III) isopropoxide acts as a precursor for the preparation of indium oxide containing films. It finds application in nano-materials and thin films display. Furthermore, it is used in imaging catalysis and in preparation of sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Indium(III) isopropoxide
145°C (decomposition)
(metals basis)
In[OCH(CH3)2]3
2 g
Moisture sensitive
Soluble in alcohols,ketones and esters.
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]
292.082
292.08
Powder
38218-24-5
White
C9H21InO3
MFCD00210318
UN3181
triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
OVZUSPADPSOQQN-UHFFFAOYSA-N
indium(3+);propan-2-olate
57346963
99.9%

Chemical Identifiers

38218-24-5
292.082
OVZUSPADPSOQQN-UHFFFAOYSA-N
57346963
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]
C9H21InO3
MFCD00210318
triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
indium(3+);propan-2-olate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : No

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only