Learn More
Indium(III) iodide, 99.998%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 316010250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
| Indium(III) iodide | |
| 25g | |
| InI3 | |
| MFCD00016151 | |
| Solubility in water: soluble | |
| [In](I)(I)I | |
| 495.53 | |
| 495.53 | |
| Trace Metal Basis |
| 13510-35-5 | |
| 22.7 to 23.5% (In) (Complexometry) | |
| InI3 | |
| indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
| RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
| triiodoindigane | |
| 83539 | |
| 99.998% | |
| Glass bottle |
Chemical Identifiers
| 13510-35-5 | |
| 495.53 | |
| RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
| 83539 | |
| [In](I)(I)I |
| InI3 | |
| MFCD00016151 | |
| indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
| triiodoindigane |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 236-839-6
RUO â Research Use Only