missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, 99.998%, (trace metal basis), Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 316010010

 View more versions of this product

Catalog No. AC316010010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
Specifications

Specifications

Indium(III) iodide
1g
InI3
MFCD00016151
Solubility in water: soluble
[In](I)(I)I
495.53
495.53
Trace Metal Basis
13510-35-5
22.7 to 23.5% (In) (Complexometry)
InI3
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
RMUKCGUDVKEQPL-UHFFFAOYSA-K
triiodoindigane
83539
99.998%
Glass bottle

Chemical Identifiers

13510-35-5
495.53
RMUKCGUDVKEQPL-UHFFFAOYSA-K
83539
[In](I)(I)I
InI3
MFCD00016151
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
triiodoindigane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-839-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only