missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) hydroxide, 99.998% (metals basis)

Catalog No. AA1185518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1185518 50 g
AA1185509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1185518 Supplier Thermo Scientific Chemicals Supplier No. 01185518
Only null left

CAS: 20661-21-6

Indium(III) hydroxide, acts as a precursor to indium(III) oxide. It is used in batteries, glasses, ceramics, chemical reagents, semiconductor materials. It is also used used for manufacturing of panel displays and demist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20661-21-6
MDL Number MFCD00011056

Specifications

Melting Point 150°C (in -H2O)
Physical Form Powder
Quantity 50 g
Solubility Information It is insoluble in water.
Formula Weight 165.84
Percent Purity 99.998%
Assay (metals basis)
Chemical Name or Material Indium(III) hydroxide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 243-947-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only