missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) hydroxide, 99.8% (metals basis)

Catalog No. AA4010009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4010009 10 g
AA4010018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4010009 Supplier Thermo Scientific Chemicals Supplier No. 04010009
Only null left

CAS: 20661-21-6

Indium(III) hydroxide is used as a precursor to indium(III)oxide. It is also applied in batteries, glasses, ceramics, chemical reagents and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20661-21-6
MDL Number MFCD00011056

Specifications

Melting Point 150°C (in -H2O)
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 165.84
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Indium(III) hydroxide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 243-947-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only