missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) fluoride, anhydrous, 99.95% (metals basis)

CAS: 7783-52-0 | F3In

$203.01 - $808.51

Chemical Identifiers

CAS 7783-52-0
Molecular Formula F3In
MDL Number MFCD00016149
Synonym indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4552506
View Documents
Thermo Scientific Chemicals
04552506
5 g
Each for $203.01
Only null left
 
AA4552514
View Documents
Thermo Scientific Chemicals
04552514
25 g
Each for $808.51
Only null left
 
Description

Description

Indium(III) fluoride is used in the preparation of non-oxide glasses. It acts as a catalyst in the reaction of trimethylsilyl cyanide (TMSCN) to aldehydes to get cyanohydrins. It play a vital role as a catalyst in organic synthesis to prepare fluorine containing 2,4,5-trisubstituted imidazoles and polysubstituted pyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-52-0
F3In
MFCD00016149
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride

Specifications

>1200°C
7783-52-0
Powder
F3In
MFCD00016149
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride
171.82
Odorless
4.39 g/mL
(metals basis)
∼1170°C
White
5 g
InF3
14,4959
Slightly soluble in water. Freely soluble in dilute acids.
99.95%
Moisture sensitive
Indium (III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 232-005-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only