missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) fluoride, anhydrous, 96%

Catalog No. AA4011809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4011809 10 g
AA4011804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4011809 Supplier Thermo Scientific Chemicals Supplier No. 04011809
Only null left

CAS: 7783-52-0

Indium(III) fluoride is employed in the synthesis of non-oxide glasses. It promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-52-0
MDL Number MFCD00016149
Synonym indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride

Specifications

Boiling Point >1200°C
Melting Point ∼1170°C
Color White
Physical Form Powder
Quantity 10 g
Merck Index 14,4959
Solubility Information Slightly soluble in water (0;04g/100mL); Freely soluble in dilute acids
Formula Weight 171.82
Percent Purity 96%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.39 g/mL
Chemical Name or Material Indium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H332-H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-005-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only