Learn More
Indium(III) chloride hydrate, 99.99% (metals basis)
CAS: 143983-91-9 | Cl3In | 221.17 g/mol
Supplier: Thermo Scientific Chemicals 01185918
Description
Indium(III) chloride hydrate is used for mild and selective cleavage of tert-butyldimethylsilyl ethers. It is a Lewis acid, and serves as a catalyst for many organic transformations. It also catalyzes the conversion of aldehydes to gem diacetates by chemoselective process. Lithium indium hydride, an in situ reducing agent, can be prepared from indium trichloride. It catalyzes reactions such as Friedel-Crafts acylations and Diels-Alder reactions. It also catalyzes Knoevenagel condensation between the barbituric acid and the aldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Indium(III) chloride hydrate | |
| White | |
| 50 g | |
| Cl3In | |
| UN3260 | |
| Soluble in water,diethyl ether,and THF. | |
| [Cl-].[Cl-].[Cl-].[In+3] | |
| 221.17 | |
| 99.99% |
| 143983-91-9 | |
| Crystalline | |
| (metals basis) | |
| MFCD00011058 | |
| 14,4958 | |
| PSCMQHVBLHHWTO-UHFFFAOYSA-K | |
| indium(3+) trichloride | |
| 221.18 (Anhydrous) | |
| Hygroscopic |
Chemical Identifiers
| 143983-91-9 | |
| 221.17 | |
| PSCMQHVBLHHWTO-UHFFFAOYSA-K | |
| [Cl-].[Cl-].[Cl-].[In+3] |
| Cl3In | |
| MFCD00011058 | |
| indium(3+) trichloride |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 233-043-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only