Learn More
Indium(III) acetate, 99.99% (metals basis)
CAS: 25114-58-3 | C2H4InO2 | 174.87 g/mol
$188.89 - $718.48
Chemical Identifiers
| CAS | 25114-58-3 |
|---|---|
| Molecular Formula | C2H4InO2 |
| Molecular Weight (g/mol) | 174.87 |
| MDL Number | MFCD00054347 |
| InChI Key | LGHOYKSQIQISBI-UHFFFAOYSA-N |
| Synonym | indium acetate, acetic acid, indium 3+ salt |
| PubChem CID | 24191761 |
| IUPAC Name | acetic acid;indium |
| SMILES | CC(=O)O.[In] |
Description
Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25114-58-3 | |
| 174.87 | |
| LGHOYKSQIQISBI-UHFFFAOYSA-N | |
| 24191761 | |
| CC(=O)O.[In] |
| C2H4InO2 | |
| MFCD00054347 | |
| indium acetate, acetic acid, indium 3+ salt | |
| acetic acid;indium |
Specifications
| 25114-58-3 | |
| 10 g | |
| C2H4InO2 | |
| indium acetate, acetic acid, indium 3+ salt | |
| LGHOYKSQIQISBI-UHFFFAOYSA-N | |
| acetic acid;indium | |
| 24191761 | |
| Hygroscopic |
| Powder | |
| 99.99% (metals basis) | |
| MFCD00054347 | |
| Soluble in water,mineral acid and acetic acid. | |
| CC(=O)O.[In] | |
| 174.87 | |
| 291.96 | |
| Indium(III) acetate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only