missing translation for 'onlineSavingsMsg'
Learn More

Indium antimonide, Electronic Grade, 99.99% (metals basis)

Catalog No. p-7080459
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8846009 10 g
AA8846018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8846009 Supplier Thermo Scientific Chemicals Supplier No. 08846009
Only null left

In semiconductor electronics, as an infrared detector and filter | CAS: 1312-41-0 | InSb | 236.578 g/mol

Indium antimonide is used in semiconductor electronics, as an infrared detector and filter. Used in thermal imaging cameras, FLIR systems, infrared homing missile guidance systems, photo-Dember emitter and in infrared astronomy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-41-0
Molecular Formula InSb
Molecular Weight (g/mol) 236.578
MDL Number MFCD00016146
InChI Key WPYVAWXEWQSOGY-UHFFFAOYSA-N
PubChem CID 3468413
IUPAC Name indiganylidynestibane
SMILES [In]#[Sb]

Specifications

Melting Point 535°C
Physical Form Pieces
Quantity 10 g
UN Number UN1549
Merck Index 14,4948
Solubility Information Insoluble in water.
Formula Weight 236.57
Percent Purity 99.99%
Grade Electronic
Odor Odorless
Density 5.75 g/mL at (m.p.)
Assay (metals basis)
Chemical Name or Material Indium antimonide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 215-192-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only