missing translation for 'onlineSavingsMsg'
Learn More

Indigotrisulfonic Acid Potassium Salt 55.0+%, TCI America™

Supplier:  TCI America I02201G

 View more versions of this product

Catalog No. I02201G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Indigotrisulfonic Acid Potassium Salt
C16H7K3N2O11S3
1 g
XOSMXDUITYWYGR-JRYLAINFSA-K
tripotassium (Z)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
6364606
≥55.0% (HPLC)
67627-18-3
MFCD00013160
potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate
[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O
616.71
616.71
Crystalline Powder

Chemical Identifiers

67627-18-3
616.71
XOSMXDUITYWYGR-JRYLAINFSA-K
6364606
[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O
C16H7K3N2O11S3
MFCD00013160
potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate
tripotassium (Z)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions