missing translation for 'onlineSavingsMsg'
Learn More

Indigo Carmine, Reagent, ACS, Spectrum™ Chemical

I3281, 860-22-0, C16H8N2O8S2Na2

Supplier:  Spectrum Chemical Mfg Cor I3281100GM

Quantity 100 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18603438


Only null left

Description

Description

Spectrum™ Chemical Indigo Carmine, Reagent, ACS , also known as 5,5′-indigodisulfonic acid sodium salt, is a pH indicator. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications

Chemical Identifiers

860-22-0
466.35
KHLVKKOJDHCJMG-QDBORUFSSA-L
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
C16H8N2Na2O8S2
MFCD00005723
disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate

Specifications

Indigo Carmine
100%
MFCD00005723
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
466.35
Amber Glass Bottle
100 g
860-22-0
C16H8N2Na2O8S2
KHLVKKOJDHCJMG-QDBORUFSSA-L
disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
ACS
Dark Violet/Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions