missing translation for 'onlineSavingsMsg'
Learn More

Indigo Carmine, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18603437
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603437 25 g Amber Glass Bottle
18603438 100 g Amber Glass Bottle
18603439 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603437 Supplier Spectrum Chemical Mfg Cor Supplier No. I328125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

I3281, 860-22-0, C16H8N2O8S2Na2

Spectrum™ Chemical Indigo Carmine, Reagent, ACS , also known as 5,5′-indigodisulfonic acid sodium salt, is a pH indicator. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

CAS Max % 100%
Grade ACS
Packaging Amber Glass Bottle
Color Dark Violet/Blue
Quantity 25 g
Chemical Name or Material Indigo Carmine