missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indigo Carmine (FD&C Blue #2), Ricca Chemical
$218.73 - $218.73
Specifications
| Chemical Name or Material | Indigo Carmine |
|---|---|
| Grade | Laboratory |
| Color | Dark Blue |
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|
RDCI005025F
|
Ricca Chemical Company
RDCI005025F1 |
Each for $218.73
|
|
||||||
Specifications
| Indigo Carmine | |
| Laboratory | |
| Dark Blue |