missing translation for 'onlineSavingsMsg'
Learn More

Indigo Carmine (Acid Blue 74), Reagents

Catalog No. 01336752
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336752 10 g
01336751 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336752 Supplier Labchem Inc Supplier No. C611400010C1
Only null left

Indigo Carmine (Acid Blue 74), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Indigo Carmine (Acid Blue 74)
CAS 860-22-0
Boiling Point 300°C
Molecular Formula C16H8N2Na2O8S2
Synonym FD and C Blue 2
Molecular Weight (g/mol) 466.3529
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.01
Color Blue
Quantity 10 g
Physical Form Solid
Show More Show Less