missing translation for 'onlineSavingsMsg'
Learn More

Indicator Solution, For Cyanide, Certified, LabChem™

Catalog No. LC155857
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC155857 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC155857 Supplier LabChem Supplier No. LC155857

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Indicator Solution
Melting Point -95°C
Name Note For Cyanide
Boiling Point 56°C
CAS 536-17-4, 67-64-1
CAS Max % 99.98, 0.02
Identification Passes Test
Packaging Amber Glass
Specific Gravity 786kg/m3
Suitability Passes Test
Linear Formula C12H12N2OS2
UN Number UN1090
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Density 786kg/m3
Color Orange to Yellow
Solubility Information Soluble in water
Quantity 125 mL
Flash Point −0.18°C
Physical Form Liquid
Formula Weight 264.37
Grade Certified
Show More Show Less