missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indanthrone 97.0+%, TCI America™
Supplier: TCI America I042925G
Specifications
| Indanthrone | |
| C28H14N2O4 | |
| UHOKSCJSTAHBSO-UHFFFAOYSA-N | |
| 2,17-diazaheptacyclo[16.12.0.0³,¹â¶.0â´,¹³.0â¶,¹¹.0¹â¹,²â¸.0²¹,²â¶]triaconta-1(30),3,6,8,10,13,15,18,21,23,25,28-dodecaene-5,12,20,27-tetrone | |
| 6690 | |
| ≥97.0% (N) | |
| 25 g |
| 81-77-6 | |
| MFCD00046964 | |
| O=C1C2=CC=CC=C2C(=O)C2=C3NC4=CC=C5C(=O)C6=CC=CC=C6C(=O)C5=C4NC3=CC=C12 | |
| 442.43 | |
| 442.43 | |
| Blue/Purple | |
| Crystal/Powder |
Chemical Identifiers
| 81-77-6 | |
| 442.43 | |
| UHOKSCJSTAHBSO-UHFFFAOYSA-N | |
| 2,17-diazaheptacyclo[16.12.0.0³,¹â¶.0â´,¹³.0â¶,¹¹.0¹â¹,²â¸.0²¹,²â¶]triaconta-1(30),3,6,8,10,13,15,18,21,23,25,28-dodecaene-5,12,20,27-tetrone |
| C28H14N2O4 | |
| MFCD00046964 | |
| 6690 | |
| O=C1C2=CC=CC=C2C(=O)C2=C3NC4=CC=C5C(=O)C6=CC=CC=C6C(=O)C5=C4NC3=CC=C12 |
Safety and Handling
EINECSNumber : (5)-2227&(5)-3311
RTECSNumber : CB8761100
TSCA : Yes