missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indacaterol maleate

Catalog No. AC464652500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464652500 250 mg
AC464650010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464652500 Supplier Thermo Scientific Chemicals Supplier No. 464652500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 753498-25-8 | C28H32N2O7 | 508.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 753498-25-8
Molecular Formula C28H32N2O7
Molecular Weight (g/mol) 508.57
MDL Number MFCD20526769,MFCD18782702
InChI Key IREJFXIHXRZFER-UHFFFAOYNA-N
IUPAC Name 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
SMILES OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1

Specifications

Melting Point 195.0°C to 197.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso
Formula Weight 508.57
Chemical Name or Material Indacaterol maleate

RUO – Research Use Only