missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Incubator 1000 Heating Module

The Incubator 1000 Heating Module is designed for temperature-controlled shaking. Instantly upgrade your platform shaker to the modular incubator system for more efficient use of limited bench space.

Supplier:  Heidolph™ 036130460

Catalog No. 13-889-007


Only null left

Description

Description

Incubator 1000 Heating Module

  • Heating capacity of 300W allows for quick temperature adjustments
  • Digital temperature settings up to 65°C and separate display for actual temperature
  • To protect your samples from thermal damages, this unit features a safety circuit to prevent overheating
  • Electrical heater comes with low-noise blower and provides accuracy of ± 2°C up to 50°C and ± 4°C over 50°C
  • Incubator 1000 Hoods sold separately
Specifications

Specifications

Heating Module
375 x 170 x 140 mm
Titramax 1000, Unimax 1010 or Promax 1020
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions