Learn More
MilliporeSigma™ Supelco™ In-Line Pressure Regulators
Description
- Adjusts pressure closer to the point of use
- Includes 1/8 in. Swagelok Nuts and Ferrules
- Maximum Input Pressure: 400 psi
Specifications
Specifications
| For Use With (Application) | Air, H2, N2, Ar, He |
| Includes | 1/8 in. Swagelok Nuts and Ferrules |
| Pressure | 0-100 psi (Outlet) |
| Description | Ultra-High Purity Version |
| Product Type | In-Line Pressure Regulator |