missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ In-Line Pressure Gauges

Catalog No. 111004862
Change view
Click to view available options
Pressure:
0 to 30 psi
0 to 60 psi
Description:
With Tee
Without Tee
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pressure Description
111004862 0 to 30 psi Without Tee
111004705 0 to 60 psi With Tee
111004863 0 to 60 psi Without Tee
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111004862 Supplier MilliporeSigma™ Supelco™ Supplier No. 20393

Install a pressure gauge in a gas delivery system to measure pressure at any given point

Specifications

For Use With (Application) Measuring Pressure
For Use With (Equipment) Gas Delivery Systems
Gauge Steel/Copper Alloy
Includes 2 in. Steel/Copper Alloy Gauge
Pressure 0 to 30 psi
Quantity 1 Ea.
Description Without Tee
Type In-Line Pressure Gauge