Learn More
Agilent Technologies™ IMT Assay (Anti-CD71) Sample Kit
Description
- Flexible assays use label free methods for more physiological assay conditions, eliminating labeling or secondary assays
- Real-time quantitative monitoring of both fast (hours) and slow (days) killing kinetics
- Sensitive assays capable of evaluating low effector cell to target cell ratios that are physiologically relevant
- Simple workflow that requires only the addition of effector cells to target cells in the presence or absence of antibodies and minimal sample handling with homogeneous assay
Specifications
Specifications
| Includes | 90 μL Tethering Reagent, 10 mL Tethering Buffer, 1.5 mL Cytolysis Reagent |
| Quantity | 1 Kit |
| Type | IMT Assay Kit |
| System Type | CellSensor™ |