missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imperial™ Protein Stain

Catalog No. PI24617
Change view
Click to view available options
Quantity:
1 L
3 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI24617 3 x 1 L
PI24615 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI24617 Supplier Thermo Scientific™ Supplier No. LSG24617

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect proteins in the low-nanogram range with this ready-to-use, in-gel, colorimetric stain formulated to deliver fast and sensitive SDS-PAGE staining.

Thermo Scientific™ Imperial Protein Stain is a unique, ready-to-use colorimetric stain formulated with coomassie dye R-250 that delivers fast and sensitive total protein staining performance for low nanogram-level detection of proteins in polyacrylamide electrophoresis gels.

Imperial Protein Stain is a coomassie dye reagent for detection of protein bands in sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE) and 2D gels. The stain is a unique formulation of coomassie brilliant blue R-250 that delivers substantial improvements in protein-staining performance compared to homemade or other commercial stains. Staining results in intensely colored protein bands that are easy to photograph and document with gel imagers. This reagent is one of the most sensitive colorimetric stains available, easily detecting 3 to 6 nanograms of protein per band. The Imperial Protein Stain protocol uses simple water-wash steps rather than methanol/acetic acid fixation and destaining, which saves valuable preparation time and minimizes reagent costs.

Highlights:

  • Sensitive – detects less than 3ng protein per band with the enhanced protocol (3 hours)
  • Fast – ready-to-use reagent detects less than 6ng protein per band in just 20 minutes
  • Robust – highly consistent, reproducible protein staining technique
  • High contrast – intense purple bands are easier to photograph or scan than typical coomassie blue stains
  • Versatile – compatible with downstream mass spectrometry analysis and protein sequencing
  • Convenient – water washes only; no acid-fixative or methanol destaining solutions required
  • Stable – 1-year room-temperature stability ensures consistent performance and saves refrigerator space
  • Flexible – adjust staining and washing times to meet time- and sensitivity-requirements

Specifications

Content And Storage Upon receipt store product at room temperature.
Detection Location In-Blot Detection, In-Gel Detection
Detection Method Colorimetric
Label or Dye Coomassie
Quantity 3 x 1 L
Target Molecule Protein
Product Line Imperial
Product Type Protein Stain
I am seeing visible aggregates in my bottle of Imperial Protein Stain. Is this normal?

Imperial Protein Stain contains additives that help slow formation of dye-dye aggregates, which occur in all Coomassie dye-based protein staining reagents. If left undisturbed, the reagent may form visible aggregates that settle in the bottle; however, gentle mixing completely disperses these aggregates. To ensure that a homogeneous sample of the reagent is used, mix reagent before pouring or dispensing.

After staining my protein gel with Imperial Protein Stain, I am not seeing any band development. What went wrong?

Here are possible causes and solutions:

- No protein was present in sample. Load a known amount of purified protein as a control.
- Insufficient amount of protein in sample. Load more total protein in gel.
- SDS not completely removed from gel. Wash gel more extensively before staining.

I stained my protein gel with Thermo Scientific Imperial Protein Stain and am getting very high background. Do you have any tips?

This is most likely due to SDS interference. We recommend washing the gel extensively before starting the staining procedure.

Is the Imperial Protein Stain compatible with mass spectrometry analysis?

Yes, here (https://tools.thermofisher.com/content/sfs/brochures/TR0050-Stained-gels-for-MS.pdf) is the procedure for processing gels stained with Imperial Stain, for mass spectrometry analysis.

What is the sensitivity of the Imperial Protein Stain?

With the enhanced protocol, the Imperial Protein Stain can detect less than 3 ng protein per band in 3 hours.

Does Imperial Protein Stain contain Coomassie G-250 or Coomassie R-250?

Imperial Protein Stain contains Coomassie R-250. The rest of the composition is proprietary.

How should I store the Thermo Scientific Imperial Protein Stain and what is its shelf life?

We recommend storing the Imperial Protein Stain at room temperature where it is stable for a year.


For Research Use Only. Not for use in diagnostic procedures.