missing translation for 'onlineSavingsMsg'
Learn More

Imperatorin 98.0+%, TCI America™

Catalog No. p-7131647
Change view
Click to view available options
Quantity:
25 mg
100 mg
Catalog No. Quantity
I090425MG 25 mg
I0904100MG 100 mg
2 options

Catalog No. I090425MG

Supplier: TCI America I090425MG

Only null left

Chemical Identifiers

CAS 482-44-0
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00016881
InChI Key OLOOJGVNMBJLLR-UHFFFAOYSA-N
Synonym 8-(3-Methyl-2-butenyloxy)psoralen, 8-Prenyloxypsoralen, Marmelosin, 9-[(3-Methyl-2-buten-1-yl)oxy]-7H-furo[3,2-g]chromen-7-one
PubChem CID 10212
ChEBI CHEBI:5885
IUPAC Name 9-(3-methylbut-2-enoxy)furo[3,2-g]chromen-7-one
SMILES CC(=CCOC1=C2C(=CC3=C1OC=C3)C=CC(=O)O2)C

Specifications

Melting Point 99°C
Quantity 25 mg
Formula Weight 270.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imperatorin
RTECSNumber LV1575000
TSCA No
Recommended Storage Freezer