Learn More
Impacto™ IMPACTOE Steel Toe Cap Overshoes
Catalog No. 19400150
Change view
Click to view available options
Size:
Men 7.5 to 8, Women: 9 to 9.5
Men: 10 to 11.5, Women: 11.5 to 13
Men: 12 to 12.5
Men: 13 to 14
Men: 5.5 to 6, Women: 7 to 7.5
Men: 6.5 to 7, Women: 8 to 8.5
Men: 8.5 to 9, Women: 10 to 11
Color:
Black
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19400150 Men: 10 to 11.5, Women: 11.5 to 13 Black
19400146 Men: 5.5 to 6, Women: 7 to 7.5 Black
19400147 Men: 6.5 to 7, Women: 8 to 8.5 Black
19400148 Men 7.5 to 8, Women: 9 to 9.5 Black
19400149 Men: 8.5 to 9, Women: 10 to 11 Black
19400151 Men: 12 to 12.5 Black
19400152 Men: 13 to 14 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19400150 Supplier Impacto™ Supplier No. IMPACTOECL
Only null left
Impacto™ Safety Toe Cap protects the toe area and is ideal for any working environment that carries a risk of accidental toe damage or has a steel toe cap requirement.
Impacto™ IMPACTOE Steel toe cap Overshoes
  • Designed to fit tightly over existing shoe
  • Made of vulcanized rubber
  • Flat anti-slip under sole for greasy and oily environment
  • Waterproof - will not harden, crack
  • Acid, oil and animal fat resistant
  • Pull-on-tab and wide shank for true slip-on entry to provide stretch
  • Recommended for use in construction and heavy work environments
  • Transferable from person to person without concern for hygiene or size restraints
  • Color-coded toe for easy identification of sizes
Specifications
Certifications/Compliance CE
Color Black
Material Vulcanized Rubber
Size Men: 10 to 11.5, Women: 11.5 to 13
Toe Material Steel
Gender Unisex
Waterproof Yes
Product Type IMPACTOE safety toe cap