Learn More
Impacto™ IMPACTOE Steel Toe Cap Overshoes
Impacto™ Safety Toe Cap protects the toe area and is ideal for any working environment that carries a risk of accidental toe damage or has a steel toe cap requirement.
Supplier: Impacto™ IMPACTOECM
Description
Impacto™ IMPACTOE Steel toe cap Overshoes
- Designed to fit tightly over existing shoe
- Made of vulcanized rubber
- Flat anti-slip under sole for greasy and oily environment
- Waterproof - will not harden, crack
- Acid, oil and animal fat resistant
- Pull-on-tab and wide shank for true slip-on entry to provide stretch
- Recommended for use in construction and heavy work environments
- Transferable from person to person without concern for hygiene or size restraints
- Color-coded toe for easy identification of sizes
Specifications
| CE | |
| Vulcanized Rubber | |
| Steel | |
| Yes |
| Black | |
| Men: 8.5 to 9, Women: 10 to 11 | |
| Unisex | |
| IMPACTOE safety toe cap |