Learn More
Impacto™ Ergomate Tracmates Anti-Slip Safety Soles
Catalog No. 19500182
Encompass
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19500182 X-Large
1 options
Catalog No. 19500182 Supplier Impacto™ Supplier No. G88602
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Impacto™ Ergomates Tracmates are used on any slippery and wet surfaces where there is risk of injury due to falling
Impacto™ Ergomate Tracmates Strap-On Soles
  • Reusable design, ideal for any person who is stripping, washing, or scrubbing floors; food processing, working on slippery surfaces; custodial and janitorial workers, hospitals, kitchens, and health care facilities
  • Not recommended for use on dry floors
  • Gain greater traction when: stripping wax floors, washing and scrubbing surfaces, on slick ceramic tiles, working on slippery concrete, walking in food processing areas, working on wet surfaces
Specifications
Color Black, Green
For Use With (Application) Stripping wax from floor, washing/scrubbing floors, slick kitchen ceramic tiles, slippery concrete, food processing areas, wet surfaces
Material EVA
Size X-Large
Toe Material Plain Toe
Gender Unisex
Product Type Safety Soles