missing translation for 'onlineSavingsMsg'
Learn More

Impacto™ Fingerless Anti-Impact Glove Liners with VEP Padding

Catalog No. p-4717643
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
Catalog No. Size Hand
19031856 Small Left
19031866 Small Right
19031855 Medium Left
19031865 Medium Right
19031853 Large Left
19031864 Large Right
19031857 X-Large Left
7 options

Catalog No. 19031856

Supplier: Impacto™ 50100120021

Only null left

Stretch polycotton fabric ensures breathability and comfort, while VEP padding protects palms from impact and shock.

  • 4-way stretch polycotton fabric to ensure breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm
  • Fingerless design for tactile feedback and mobility
  • Designed to be worn under work gloves for an additional level of protection
  • Ideal for protection from electric, manual, and power tools
  • Sold in pairs

Specifications

Certifications/Compliance (Europe) EN 420: 2003 + A1:2009
Color Blue
Hand Left
Material Polycotton
Product Type Glove Liners
Size Small
EN388 Standard EN388:3231
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Palm Type VEP Padding
Series 501-00
Show More Show Less