missing translation for 'onlineSavingsMsg'
Learn More

Impacto™ Fingerless Anti-Impact Glove Liners with VEP Padding

Catalog No. 19031856
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Hand:
Left
Right
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19031856 Small Left
19031866 Small Right
19031855 Medium Left
19031865 Medium Right
19031853 Large Left
19031864 Large Right
19031857 X-Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19031856 Supplier Impacto™ Supplier No. 50100120021

Stretch polycotton fabric ensures breathability and comfort, while VEP padding protects palms from impact and shock.

  • 4-way stretch polycotton fabric to ensure breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm
  • Fingerless design for tactile feedback and mobility
  • Designed to be worn under work gloves for an additional level of protection
  • Ideal for protection from electric, manual, and power tools
  • Sold in pairs

Specifications

Certifications/Compliance (Europe) EN 420: 2003 + A1:2009
Color Blue
Hand Left
Material Polycotton
Product Type Glove Liners
Size Small
EN388 Standard EN388:3231
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Palm Type VEP Padding
Series 501-00
Show More Show Less