missing translation for 'onlineSavingsMsg'
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Suede Leather Palms

Catalog No. p-3668032
Change view
Click to view available options
Size:
Small
Medium
Hand:
Left
Right
Catalog No. Size Hand
19031893 Small Right
19031874 Medium Left
19031892 Medium Right
3 options

Catalog No. 19031893

Supplier: Impacto™ 50110110022

Only null left

Nylon fabric offers breathability and comfort, while suede leather on the palms protects from abrasion.

  • Stretch Nylon Lycra™ material offers breathability and comfort
  • Suede leather offers abrasion protection and increased gripping abilities
  • Contoured VEP 1/8 in. padding in the palm provides protection from impact and shock
  • Fingerless design for tactile feedback and mobility

Specifications

Back Material Nylon Lycra™
Coating Material Uncoated
Color Blue, Gray
Hand Right
Material Suede
Product Type Anti-impact Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Machinery/Power Tool Operation
Glove Cuff Style Open
Series 501-10
Show More Show Less