Learn More
Impacto™ Fingerless Grain Leather Anti-Vibration Gloves with Detachable Wrist Support
Catalog No. 19031952
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
Hand:
Left
Right
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19031952 Small Right
19031930 Small Left
19031950 Medium Right
19031948 Large Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19031952 Supplier Impacto™ Supplier No. 70420120022
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fingerless gloves feature VEP padding in the palm and thumb web area for protection against abrasion, impact, and shock. Detachable wrist support minimizes extension and over-flexion of the wrist to prevent repetitive strain injuries (RSI).
  • VEP padding protects the palm and thumb web area from impact
  • Anti-shock wrist support protects from Repetitive Strain Injuries (RSI) such as Carpal Tunnel Syndrome or Tendinitis
  • Wrist support minimizes extension and over flexion of the wrist
  • Provides protection from abrasion, impact, and shock
  • Fingerless style for optimum dexterity
  • Use with hand or electric tools in repetitive tasks
Specifications
Cuff Material Grain Leather
Color Blue, Yellow
Hand Right
Material Nylon, Grain Leather
Product Type Fingerless Gloves
Size Small
Finger Style Fingerless
Glove Cuff Color Yellow
Series 704-20