Learn More
Impacto™ Fingerless Grain Leather Anti-Vibration Gloves with Detachable Wrist Support
Fingerless gloves feature VEP padding in the palm and thumb web area for protection against abrasion, impact, and shock. Detachable wrist support minimizes extension and over-flexion of the wrist to prevent repetitive strain injuries (RSI).
Supplier: Impacto™ 70420120042
Description
- VEP padding protects the palm and thumb web area from impact
- Anti-shock wrist support protects from Repetitive Strain Injuries (RSI) such as Carpal Tunnel Syndrome or Tendinitis
- Wrist support minimizes extension and over flexion of the wrist
- Provides protection from abrasion, impact, and shock
- Fingerless style for optimum dexterity
- Use with hand or electric tools in repetitive tasks
Specifications
| Grain Leather | |
| Right | |
| Fingerless Gloves | |
| Fingerless | |
| 704-20 |
| Blue, Yellow | |
| Nylon, Grain Leather | |
| Large | |
| Yellow |