missing translation for 'onlineSavingsMsg'
Learn More

Impacto™ Cush'N Step Anti-Fatigue Flat Insoles

Catalog No. 19039341
Change view
Click to view available options
Size:
11 to 12.5
9 to 10.5
Gender:
Men's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Gender
19039341 9 to 10.5 Men's
19039347 11 to 12.5 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039341 Supplier Impacto™ Supplier No. CUSHFLATD F

Impacto™ Cush'N Step CUSHFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

Impacto™ Cush'N Step Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes

Specifications

Color Green
For Use With (Application) Use with dress and low cut footwear
Length (English) 9 in.
Length (Metric) 22.86 cm
Material Open-cell Foam
Size 9 to 10.5
Style Flat
Gender Men's
Product Type Full-tapered Insole