missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products GL201 Heavy Duty Anti-Vibration Mechanics Gloves with TPR Padding

Catalog No. 19555045
Change view
Click to view available options
Size:
2X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19555045 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19555045 Supplier Pyramex Safety Products Supplier No. GL201X2
Only null left

Gloves feature TPR padding on three fingers for protection against impacts, and anti-shock EVA padding on the index finger.

  • Two-layer stretch spandex construction for comfort and flexibility
  • TPR padding on three fingers for impact protection
  • Anti-shock EVA padding on the index finger and palm
  • PVC patches on the palm for enhanced abrasion resistance
  • Soft terry cloth brow-wipe on the back of the glove
  • Hook and loop closure

Specifications

Back Material Spandex
Coating Material Uncoated
Color Black, Blue
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size 2X-Large
EN388 Standard 2121
Finger Style Full
For Use With (Application) Carpentry, Construction, Machine Operation, and Mechanics
Glove Cuff Style Hook and Loop
Palm Type PVC Patches with EVA Padding
Series GL201
Show More Show Less