Learn More
Stemcell Technologies Inc ImmunoCult™-ACF Dendritic Cell Medium, Size: 100 mL
Catalog No. 501974485
Encompass
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation
Catalog No. 501974485 Supplier Stemcell Technologies Inc Supplier No. 10987
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation