missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Standard Modules White and Black

Catalog No. 12565289A
Change view
Click to view available options
Color:
Black
White
Volume (Metric) Well:
350 μL
400 μL
Surface Treatment:
MaxiSorp
PolySorp
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Well Surface Treatment
12565289A White 350 μL MaxiSorp
12565289B White 350 μL PolySorp
12565274 Black 400 μL MaxiSorp
12565275 Black 400 μL PolySorp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12565289A Supplier Thermo Scientific™ Supplier No. 437591

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.

Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

Specifications

Breakapart No
Binding Property Hydrophilic
Certified Yes
Format Standard
Lid Without Lid
Detection Method Chemiluminescence
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Color White
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 250 μL
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
For Use With (Application) Fluorescence and Luminescence
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Show More Show Less