missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Supplier:  Thermo Scientific™ 467466

 View more versions of this product

Catalog No. 12565119


Only null left

Description

Description

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Certifications

Products specified in chart are certified for: Adsorption of IgG; CV of <5% between wells; all results +/-10% from mean; and background of all wells +/-0.005 absorbance units from mean.

Specifications

Chemical Identifiers

Colorimetric
Clear
None

Specifications

No
Certified for adsorption of IgG; CV of <5% between wells.
Strip Plate
Colorimetric
60 Cases
Strip Plate
96
Non-sterile
Round, Flat Bottom (F-well)
10
60/Cs.
F16 (2x8 wells)
Hydrophilic
Yes
Without Lid
8 x 12
Clear
Polystyrene
350 μL
MaxiSorp
60
None
Antibodies (Glycoproteins)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions