Learn More
Thermo Scientific™ Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Supplier: Thermo Scientific™ 95029390
Description
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
Features:
- For solid-phase immunoassays, e.g. ELISA
- Choice of four surfaces for optimal binding
- Free-standing modules are easy to handle
- Alphanumeric system for easy orientation
- Fits standard equipment
- High optical quality for reliable results
- Polystyrene
Choice of Surface:
- PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
- MediSorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
- MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
- MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins
Chemical Identifiers
| Strip Plate | |
| Colorimetric | |
| Polystyrene | |
| None |
| High optical quality for reliable results | |
| Clear | |
| 96 |
Specifications
| Breakable Strips | |
| No | |
| High optical quality for reliable results | |
| 25 Pk., 50 Cases | |
| Clear | |
| Polystyrene | |
| 96 | |
| Universal Binding | |
| For adsorption of hydrophobic molecules | |
| 50 | |
| Lipids |
| Hydrophobic | |
| Strip Plate | |
| Colorimetric | |
| Adsorption | |
| Microplate Strip | |
| 400 μL | |
| 350 μL | |
| Round, Flat Bottom (F-well) | |
| 50 | |
| None |