Learn More
Thermo Scientific™ Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Supplier:  Thermo Scientific™ 446490
 View more versions of this product
Catalog No. 1496220
Only null left
Description
Description
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
Features:
  • For solid-phase immunoassays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fits standard equipment
  • High optical quality for reliable results
  • Polystyrene
Choice of Surface:
  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • MediSorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins
Specifications
Chemical Identifiers
Strip Plate
Colorimetric
Polystyrene
None
High optical quality for reliable results
Clear
96
Specifications
Breakable Strips
No
Without Lid
Colorimetric
10 Pk.
Frame
350 μL
250 μL
Round, C Bottom
60
None
Very Hydrophilic
Strip Plate
High optical quality for reliable results
8 x 12
Clear
Polystyrene
96
MultiSorp
For glycans and water soluble proteins
10
Glycans
Product Suggestions
Product Suggestions