missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Breakable Modules Clear, C8 LockWell, MediSorp

Catalog No. 12565732
Change view
Click to view available options
Surface Treatment:
MediSorp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
12565732 MediSorp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565732 Supplier Thermo Scientific™ Supplier No. 446470

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.

Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
  • For solid phase immuno assays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fit standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Specifications

Breakapart Breakable Strips
Binding Property Slightly Hydrophilic
Certified Yes
Lid Without Lid
Description High optical quality for reliable results
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Frame
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Surface Treatment MediSorp
Well Shape Round, Curved Edges, Flat Bottom (C-well)
For Use With (Application) For binding a diverse range of biomolecules
No. per Case 60
No. per Pack 10
Plate Blocking None
Target Molecule Proteins
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?

Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.