missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Immune System Education Kit

Catalog No. S001198
Change view
Click to view available options
Subject:
Biology
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Subject
S001198 Biology
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001198 Supplier United Scientific™ Supplier No. IMMUNEKIT

missing translation for 'validMainframeMsg'

Prop 65

Kit is designed to aid in teaching the fundamentals of the immune system. Students can investigate how the body defends itself against antigens and viruses.

  • Kit is designed to aid in teaching the fundamentals of the immune system.
  • Students can investigate how the body defends itself against antigens and viruses.
  • Understanding that students learn best when information is presented orally, visually, and tactilely teachers are provided with animated slides to project on a wall or screen.
  • 50 custom plastic models representing immune system cells, proteins and key structures help simulate how the immune system works.
  • Student assessment can be evaluated via activities utilizing kit models and follow-up questions.
  • Kit contains no consumables and can be shared among teachers.
  • The durability of the plastic models will provide for years of use. Includes downloadable teacher resources.
  • Features hands-on models to investigate immune system functions
  • Includes 50 custom plastic models of immune system cells and proteins
  • Includes downloadable teacher resources

Specifications

Grade 7 to 12
Format Kit
Product Type Immune System Education Kit
Languages English
Subject Biology