Learn More
Vector Laboratories ImmPACT DAB EqV Peroxidase (HRP) Substrate | 100mL
Catalog No. NC1883496
Encompass
ImmPACT DAB EqV consists of equal volumes of Reagent 1 and Reagent 2, and when mixed together form a working solution. ImmPACT DAB EqV produces a dark brown reaction product and is 3 to 5 times more sensitive than the original DAB substrate.
Catalog No. NC1883496 Supplier Vector Laboratories Supplier No. SK4103100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ImmPACT DAB EqV consists of equal volumes of Reagent 1 and Reagent 2, and when mixed together form a working solution. ImmPACT DAB EqV produces a dark brown reaction product and is 3 to 5 times more sensitive than the original DAB substrate.