missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod 98.0+%, TCI America™

Catalog No. I0747100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0747100MG 100 mg
I07471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0747100MG Supplier TCI America Supplier No. I0747100MG

Chemical Identifiers

CAS 99011-02-6
Molecular Formula C14H16N4
Molecular Weight (g/mol) 240.31
MDL Number MFCD00866946
InChI Key DOUYETYNHWVLEO-UHFFFAOYSA-N
Synonym imiquimod, aldara, zyclara, beselna, 1-2-methylpropyl-1h-imidazo 4,5-c quinolin-4-amine, zartra, imiquimod acetate, 1-isobutyl-1h-imidazo 4,5-c quinolin-4-amine, 4-amino-1-isobutyl-1h-imidazo 4,5-c quinoline
PubChem CID 57469
ChEBI CHEBI:36704
IUPAC Name 1-(2-methylpropyl)-1H-imidazo[4,5-c]quinolin-4-amine
SMILES CC(C)CN1C=NC2=C1C1=CC=CC=C1N=C2N

Specifications

Melting Point 294°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 240.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Imiquimod
RTECSNumber NJ5903450
TSCA No