missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, 99%

Catalog No. AAJ63990MC
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63990MC 100 mg
AAJ6399003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63990MC Supplier Thermo Scientific Chemicals Supplier No. J63990MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99011-02-6 | C14H16N4 | 240.31 g/mol

Imiquimod is a caspase 3 activator which acts as an immunomodulator and displays antiviral and anti-tumor activity. It is a patient-applied cream used for the treatment of genital warts and basal cell carcinoma. It is also used to cure actinic keratosis on the face and scalp. It belongs to a group of drugs called immune response modifiers, which work by activating the immune system to fight abnormal skin growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide. Slightly soluble in water. Insoluble in ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 99011-02-6
Molecular Formula C14H16N4
Molecular Weight (g/mol) 240.31
MDL Number MFCD00866946
InChI Key DOUYETYNHWVLEO-UHFFFAOYSA-N
Synonym imiquimod, aldara, zyclara, beselna, 1-2-methylpropyl-1h-imidazo 4,5-c quinolin-4-amine, zartra, imiquimod acetate, 1-isobutyl-1h-imidazo 4,5-c quinolin-4-amine, 4-amino-1-isobutyl-1h-imidazo 4,5-c quinoline
PubChem CID 57469
ChEBI CHEBI:36704
IUPAC Name 1-(2-methylpropyl)imidazo[4,5-c]quinolin-4-amine
SMILES CC(C)CN1C=NC2=C1C1=CC=CC=C1N=C2N

Specifications

Melting Point 292°C to 294°C
Color White
Quantity 100 mg
UN Number UN2811
Merck Index 144922
Solubility Information Soluble in dimethyl sulfoxide and dimethyl formamide; Slightly soluble in water; Insoluble in ethanol.
Formula Weight 240.31
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Imiquimod
Hazard Category Acute toxicity (category 2), Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Fatal if swallowed, Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only