missing translation for 'onlineSavingsMsg'
Learn More

Imipenem monohydrate, 95%

Catalog No. p-7122684
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66749LB0 5 mg
AAJ66749MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66749LB0 Supplier Thermo Scientific Chemicals Supplier No. J66749LB0

Imipenem monohydrate,Tienam, Primaxin, CAS # 74431-23-5 is a broad-spectrum carbapenem antibiotic that interferes with the activity of penicillin binding proteins along the inner cell wall. | CAS: 74431-23-5 | C12H19N3O5S | 317.36 g/mol

Imipenem monohydrate is a broad spectrum B-lactam antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
  • Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
  • Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria

Applications

  • Suitable for the study of antibiotic resistance
  • High-quality, laboratory grade for use in vitro experiments

Chemical Identifiers

CAS 74431-23-5
Molecular Formula C12H19N3O5S
Molecular Weight (g/mol) 317.36
MDL Number MFCD09753321
InChI Key GSOSVVULSKVSLQ-XMCAPODCSA-N
PubChem CID 5282372
ChEBI CHEBI:51799
IUPAC Name (5R,6S)-3-({2-[(aminomethylidene)amino]ethyl}sulfanyl)-6-[(1S)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
SMILES O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O

Specifications

Type Imipenem Monohydrate
Physical Form Powder
Melting Point 193°C to 198°C
Color White
Quantity 5 mg
Flash Point >110°C (230°F)
Boiling Point 567°C
Merck Index 14,4919
Synonym Primaxin; Tienam
Solubility Information Soluble in water at 5mg/mL
Formula Weight 317.36
Percent Purity 95%
Chemical Name or Material Imipenem monohydrate
Show More Show Less
EINECSNumber 264-734-5
RTECSNumber CL5446516
TSCA No
storageNote1 −30° to −10°C
Recommended Storage Store at -20°C

RUO – Research Use Only