Learn More
Iminostilbene, 97%
CAS: 256-96-2 | C14H11N | 193.249 g/mol
Supplier: Thermo Scientific Chemicals B2132314
Description
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.
Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Iminostilbene | |
| 197°C to 199°C | |
| MFCD00005071 | |
| 1343358 | |
| Soluble in ethyl acetate (25mg/ml),water (partly),dioxane,chloroform,and DMSO. | |
| C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2 | |
| 193.249 | |
| CHEBI:47802 | |
| 97% |
| 256-96-2 | |
| C14H11N | |
| 25 g | |
| iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino | |
| LCGTWRLJTMHIQZ-UHFFFAOYSA-N | |
| 11H-benzo[b][1]benzazepine | |
| 9212 | |
| 193.25 |
Chemical Identifiers
| 256-96-2 | |
| 193.249 | |
| LCGTWRLJTMHIQZ-UHFFFAOYSA-N | |
| 9212 | |
| 11H-benzo[b][1]benzazepine |
| C14H11N | |
| MFCD00005071 | |
| iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino | |
| CHEBI:47802 | |
| C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2 |
Safety and Handling
EINECSNumber : 205-970-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only